Learn More
New Pig HazMat Quick Response High Visibility Spill Cart
SDP
Catalog No. 18813363 Shop All New Pig Corporation Products
Change view
Click to view available options
Material:
Polypropylene
Catalog No. Material
18-813-363 Polypropylene
1 options
Catalog No. 18-813-363 Supplier New Pig Supplier No. KIT3401
Add to Cart
Add to Cart
Each absorbs up to 20 gallons, absorbs acids, bases, unknowns.
  • 20 gallons of easily accessible HazMat absorbents and products packed neatly inside
  • Wheeled, yellow, high-visibility HazMat spill kit.
  • Respond to small and moderate sized HazMat spills
  • Lockable front-loading doors open wide in a HazMat emergency
Specifications
Material Polypropylene
Type Spill Control Cart
Weight Capacity Heavyweight
Weight (English) 62 lb.
Weight (Metric) 28.1 kg