Learn More
New Pig HazMat Spill Kit in 20 Gallon Overpack Salvage Drum
SDP
Catalog No. 18813364 Shop All New Pig Corporation Products
Change view
Click to view available options
Material:
Polypropylene
Catalog No. Material
18-813-364 Polypropylene
1 options
Catalog No. 18-813-364 Supplier New Pig Supplier No. KIT311
Add to Cart
Add to Cart
Each absorbs up to 11 gal., absorbs acids, bases, unknowns.
  • 20 gallon overpack salvage drum
  • Absorbs up to 11 gal.
  • Absorbs acids, bases, and unknowns
Specifications
Material Polypropylene
Type Drum Spill Kit
Weight Capacity Heavyweight
Weight (English) 27.9 lb.
Weight (Metric) 12.7 kg