Learn More
New Pig HazMat Spill Kit in High-Visibility Bag
SDP
Catalog No. 18389123 Shop All New Pig Corporation Products
Change view
Click to view available options
Type: HazMat Spill Kit in High-Visibility Bag
Catalog No.
18-389-123 Type: HazMat Spill Kit in High-Visibility Bag
1 options
This item is not returnable. View return policy
Catalog No. 18-389-123 Supplier New Pig Supplier No. KIT320
  • $383.00
    $362.65 / Each of 1
    Save $20.35
    5% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
This item is not returnable. View return policy
Chemical absorbents in an easy to see, fast to find, high-visibility bag.
  • Durable, water-resistant bag with hook-and-loop closure keeps contents clean and dry
  • PIG HazMat Socks stop spreading spills; PIG HazMat Pads absorb quickly
  • PIG HazMat Absorbents are specially treated for unsurpassed performance with concentrated corrosives, such as 98% sulfuric acid and 30% sodium hydroxide
  • Temporary disposal bags help make cleanup easier
  • Bright yellow bag and large, color-coded label make the kit easy to locate
  • Strap for easy carrying and hanging
  • Compact and easy to store - makes a great truck spill kit
Specifications
Material Polypropylene