Learn More
Description
This antibody detects a major band around 14-16 kDa in the western blot analysis of heart tissue and K-562 cells. A higher band around 26-27 kDa can also be observed occasionally, which may represents the dimer form of HBA1. The antibody is specific to HB
Specifications
Specifications
| Antigen | HBA1 |
| Applications | Immunocytochemistry, Immunofluorescence, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.24 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| Gene | HBA1 |
| Gene Accession No. | P69905 |
| Gene Alias | Alpha globin, HBA1, HBA2, Hemoglobin alpha chain, Hemoglobin subunit alpha, hemoglobin, alpha 1 |
| Gene Symbols | HBA1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
