Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HBEGF Monoclonal Antibody (D2)
GREENER_CHOICE

Catalog No. MA566067
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA566067 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA566067 Supplier Invitrogen™ Supplier No. MA566067
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HBEGF Monoclonal Antibody for Western Blot

HBEGF is a growth factor that mediates its effects via EGFR, ERBB2 and ERBB4. It is required for normal cardiac valve formation and normal heart function. HBEGF promotes smooth muscle cell proliferation. It may be involved in macrophage-mediated cellular proliferation. It is mitogenic for fibroblasts, but not endothelial cells. It is able to bind EGF receptor/EGFR with higher affinity than EGF itself and is a far more potent mitogen for smooth muscle cells than EGF. It also acts as a diphtheria toxin receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HBEGF
Applications Western Blot
Classification Monoclonal
Clone D2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene HBEGF
Gene Accession No. Q99075
Gene Alias AW047313; Diphtheria toxin receptor; Diphtheria toxin receptor (heparin binding epidermal growth factor - like growth factor); diphtheria toxin receptor (heparin-binding EGF-like growth factor); diphtheria toxin receptor (heparin-binding epidermal growth factor-like growth factor); DTR; DT-R; DTS; DTSF; GFHB; hb egf; HBEGF; Hb-egf; Hegfl; heparin binding EGF like growth factor; heparin binding epidermal growth factor-like growth factor; heparin-binding EGF-like growth factor; heparin-binding epidermal -growth - like growth factor; heparin-binding epidermal growth factor; heparin-binding epidermal -growth factor; proheparin-binding EGF-like growth factor
Gene Symbols HBEGF
Host Species Mouse
Immunogen Recombinant Heparin Binding Epidermal Growth Factor Like Growth Factor (HBEGF) (Val21-Thr160).
Purification Method Protein A/G
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1839
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.