Learn More
Cytiva HBS-EP+ Buffer 10, 1000mL
Catalog No. 501055344 Shop All Cytiva Products
Encompass
General purpose running buffer. Concentrated stock solution yielding pH 7.4 when diluted 10x. For use in Biacore 4000, Biacore T200, Biacore T100, Biacore 8K, Biacore S200 and Biacore X100
Catalog No. 50-105-5344 Supplier Cytiva Supplier No. BR100669
General purpose running buffer. Concentrated stock solution yielding pH 7.4 when diluted 10x. For use in Biacore 4000, Biacore T200, Biacore T100, Biacore 8K, Biacore S200 and Biacore X100