Learn More
Cytiva HBS-EP+ Buffer 10, 4x50mL
Catalog No. 501055354 Shop All Cytiva Products
Encompass
General purpose running buffer. Concentrated stock solution yielding pH 7.4 when diluted 10x. For use in Biacore 4000, Biacore T200, Biacore T100, Biacore 8K, Biacore S200 and Biacore X100
Catalog No. 50-105-5354 Supplier Cytiva Supplier No. BR100826
General purpose running buffer. Concentrated stock solution yielding pH 7.4 when diluted 10x. For use in Biacore 4000, Biacore T200, Biacore T100, Biacore 8K, Biacore S200 and Biacore X100