Learn More
Cytiva HBS-EP BUFFER,READY-TO-USE
Supplier:  Cytiva BR100188
Encompass
General purpose buffer, degassed and ready to use 0.01 M HEPES pH 7.4, 0.15 M NaCl, 3 mM EDTA, 0.005% v/v Surfactant P20
Catalog No. 50-105-5281
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.