Learn More
Cytiva HBS-N Buffer 10, 1000mL
Catalog No. 501055345 Shop All Cytiva Products
Encompass
Contents: 0.1 M HEPES, 1.5 M NaCl; Running buffer. Concentrated stock solution, yielding pH 7.4 when diluted 10X. For use in all Biacore SPR systems.
Catalog No. 50-105-5345 Supplier Cytiva Supplier No. BR100670
Contents: 0.1 M HEPES, 1.5 M NaCl; Running buffer. Concentrated stock solution, yielding pH 7.4 when diluted 10X. For use in all Biacore SPR systems.