Learn More
Cytiva HBS-N Buffer 10, 4x50mL
Catalog No. 501055356 Shop All Cytiva Products
Encompass
Contents: 0.1 M HEPES, 1.5 M NaCl; Running buffer. Concentrated stock solution, yielding pH 7.4 when diluted 10X. For use in all Biacore SPR systems.
Catalog No. 50-105-5356 Supplier Cytiva Supplier No. BR100828
Contents: 0.1 M HEPES, 1.5 M NaCl; Running buffer. Concentrated stock solution, yielding pH 7.4 when diluted 10X. For use in all Biacore SPR systems.