Learn More
Cytiva HBS-N Buffer, 6x200mL
Catalog No. 501055304 Shop All Cytiva Products
Encompass
Degassed and ready to use 0.01 M HEPES pH 7.4, 0.15 M NaCl. For use in Biacore C, and Biacore 3000 SPR systems
Catalog No. 50-105-5304 Supplier Cytiva Supplier No. BR100369
Degassed and ready to use 0.01 M HEPES pH 7.4, 0.15 M NaCl. For use in Biacore C, and Biacore 3000 SPR systems