Learn More
Cytiva HBS-P+ Buffer 10, 1000mL
Catalog No. 501055346 Shop All Cytiva Products
Encompass
Concentrated stock solution containing 0.1 M HEPES, 1.5 M NaCl and 0.5% v/v Surfactant P20. Will yield pH 7.4 when diluted 10X. Supports the recommendations for small molecule assays in Biacore systems. For all systems.
Catalog No. 50-105-5346 Supplier Cytiva Supplier No. BR100671
Concentrated stock solution containing 0.1 M HEPES, 1.5 M NaCl and 0.5% v/v Surfactant P20. Will yield pH 7.4 when diluted 10X. Supports the recommendations for small molecule assays in Biacore systems. For all systems.