Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
- Home
- Products
- Research and Clinical Diagnostic Tests and Controls
- Chemistry Controls, Calibrators and Reagents
- General Microbiology Quality Control Products
- ZeptoMetrix HBV Seroconversion Panels - 11 to 20 Member Panel
ZeptoMetrix HBV Seroconversion Panels - 11 to 20 Member Panel
Catalog No. 22157158
Shop All ZeptoMetrix Corporation Products

Change view
Click to view available options
Quantity:
11 x 1 mL
12 x 1 mL
13 x 1 mL
14 x 1 mL
15 x 1 mL
16 x 1 mL
17 x 1 mL
18 x 1 mL
19 x 1 mL
20 x 1 mL
Specimen Type:
Donor No.: 62347
Donor No.: 62433
Donor No.: 62675
Donor No.: 62825
Donor No.: 62967
Donor No.: 63253
Donor No.: 63291
Donor No.: 63997
Donor No.: 64006
Donor No.: 65099
Donor No.: 65777
Donor No.: 66201
Donor No.: 66213
Donor No.: 66481
Donor No.: 66537
Donor No.: 67303
Donor No.: 67457
Donor No.: 67694
Donor No.: 68029
Donor No.: 68105
Donor No.: 68739
Donor No.: 69751
Donor No.: 70292
Donor No.: 70793
Donor No.: 70970
Donor No.: 71415
Donor No.: 71922
Donor No.: 72439
Donor No.: 72474
Donor No.: 73299
30 product options available for selection
| Catalog No. | Quantity | Specimen Type |
|---|---|---|
| 22-157-158 | 19 x 1 mL | Donor No.: 62347 |
| 22-156-974 | 14 x 1 mL | Donor No.: 66537 |
| 22-156-973 | 17 x 1 mL | Donor No.: 66201 |
| 22-156-972 | 14 x 1 mL | Donor No.: 65777 |
| 22-156-975 | 18 x 1 mL | Donor No.: 67303 |
| 22-156-977 | 18 x 1 mL | Donor No.: 67457 |
| 22-156-978 | 14 x 1 mL | Donor No.: 67694 |
| 22-156-981 | 12 x 1 mL | Donor No.: 68029 |
| 22-156-982 | 14 x 1 mL | Donor No.: 68105 |
| 22-156-984 | 14 x 1 mL | Donor No.: 68739 |
| 22-156-986 | 16 x 1 mL | Donor No.: 70970 |
| 22-156-985 | 14 x 1 mL | Donor No.: 70793 |
| 22-156-987 | 14 x 1 mL | Donor No.: 71415 |
| 22-156-993 | 11 x 1 mL | Donor No.: 72474 |
| 22-156-990 | 16 x 1 mL | Donor No.: 70292 |
| 22-156-989 | 13 x 1 mL | Donor No.: 69751 |
| 22-156-992 | 13 x 1 mL | Donor No.: 72439 |
| 22-156-996 | 12 x 1 mL | Donor No.: 71922 |
| 22-156-998 | 15 x 1 mL | Donor No.: 73299 |
| 22-157-151 | 11 x 1 mL | Donor No.: 63291 |
| 22-157-156 | 14 x 1 mL | Donor No.: 62675 |
| 22-157-155 | 12 x 1 mL | Donor No.: 62433 |
| 22-157-157 | 11 x 1 mL | Donor No.: 62825 |
| 22-157-159 | 16 x 1 mL | Donor No.: 62967 |
| 22-157-161 | 11 x 1 mL | Donor No.: 63253 |
| 22-157-164 | 12 x 1 mL | Donor No.: 63997 |
| 22-157-166 | 12 x 1 mL | Donor No.: 64006 |
| 22-157-170 | 20 x 1 mL | Donor No.: 66213 |
| 22-157-169 | 16 x 1 mL | Donor No.: 65099 |
| 22-157-168 | 18 x 1 mL | Donor No.: 66481 |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 30 options available.
Specifications
Specifications
| Certifications/Compliance | FDA |
| Content And Storage | -20°C |
| Product Type | Seroconversion Reference Panel |
| Quantity | 19 x 1 mL |
| Specimen Type | Donor No.: 62347 |
Product Title
Select an issue
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?