Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HBZ Monoclonal Antibody (GT1544)
GREENER_CHOICE

Catalog No. PIMA527754 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527754 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527754 Supplier Invitrogen™ Supplier No. MA527754
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

Positive Control: K562 Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a member of a subfamily of lipid trafficking proteins that are characterized by a C-terminal steroidogenic acute regulatory domain and an N-terminal metastatic lymph node 64 domain. The encoded protein localizes to the membranes of late endosomes and may be involved in exporting cholesterol. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HBZ
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone GT1544
Concentration 1.54 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Hbz
Gene Accession No. P02008
Gene Alias AI450015; alpha-like embryonic globin chain x; Hba-x; HBAZ; HBZ; HBZ1; HBZ2; HBZ-T1; hemaglobin zeta; hemoglobin subunit zeta; hemoglobin X, alpha-like embryonic chain in Hba complex; Hemoglobin zeta; hemoglobin zeta chain; hemoglobin, zeta; RA_M008_JSM295ECF; RGD1307486; zeta globin; zeta-globin
Gene Symbols Hbz
Host Species Mouse
Immunogen Recombinant protein encompassing a sequence within the center region of human Hemoglobin zeta. The exact sequence is proprietary.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3050
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.