Learn More
TARGETMOL CHEMICALS INC HC-030031 100MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T6530
Encompass_Preferred
Also available in 10 mg 50 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. HC-030031 (TOSLAB 829227) is a potent TRPA1 inhibitor which antagonizes AITC- and formalin-evoked calcium influx with IC50s of 6.2±0.2 and 5.3±0.2 uM respectively. purity: 99%
Catalog No. 50-257-9339
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.