Learn More
Hourglass International™ HCE™ 2-Ply Cleanroom Face Masks with Earloops
Cleanroom face masks provide long-wear comfort while protecting the environment from human contaminants including skin flakes, cosmetics, bacteria and other particles. Suitable for Class 100/ISO 5 controlled environments.
Supplier:  Hourglass International™ FM2ESC14323
 View more versions of this product
Catalog No. 19-423-087
This item is not returnable. View return policy
Description
Description
  • Low lint design for critical environments
  • 2-ply, white, non-woven ES (PP/PE)/ meltblown polypropylene (BMPP) material
  • Cleanroom manufactured and packed
  • Double bagged cleanroom packaging
  • Soft flexible nose piece
  • Statistical manufacturing process controls
  • OP earloops attached to mask body for strength
Specifications
Specifications
Cleanroom Face Mask
Earloops
X-Large
Pleated
2-ply
Semiconductor, Disk Drive, Medical Device, Pharmaceutical, Biomedical
9 in.
FM2ESC143-23
Double-bagged, Cleanroom Packaged
White
Polyethylene
Non-sterile
Class 100 (ISO 5)
Single-use
Yes
23 cm
50/Bag, 2000/Cs.
HCE™
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.