Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

hCG alpha Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. p-7210767 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-4669 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-4669 Supplier Proteintech Group Inc Supplier No. 250141AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Luteinizing hormone (LH, also known as lutropin) is a tropic hormone which modulates the secretory activity of other endocrine glands. It is produced in the anterior hypophysis of the pituitary gland. The glycoprotein hormone, LH, like follicle stimulating hormone and thyroid stimulating hormone, is composed of a common alpha-subunit but also a specific beta-subunit, which characterizes each of these hormones.

Specifications

Antigen hCG alpha
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.36 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene CGA
Gene Accession No. P01215
Gene Alias CG ALPHA, CGA, Choriogonadotropin alpha chain, Follitropin alpha chain, FSH alpha, FSHA, GPHa, GPHA1, HCG, hCG alpha, LHA, LSH alpha, Lutropin alpha chain, Thyrotropin alpha chain, TSH alpha, TSHA
Gene Symbols CGA
Host Species Rabbit
Immunogen hCG alpha Fusion Protein Ag17313
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1081
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.