Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

hCG Antibody (1270D) [DyLight 350], R&D Systems™
SDP

Catalog No. FAB97702D
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB97702D 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FAB97702D Supplier R&D Systems Supplier No. FAB97702D

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Rabbit Monoclonal Antibody

hCG Monoclonal antibody specifically detects hCG in Human samples. It is validated for ELISA

Specifications

Antigen hCG
Applications ELISA
Classification Monoclonal
Clone 1270D
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias CGB, CGB5, CGB7, CGB8, CG-beta, Choriogonadotropin subunit beta 3, Chorionic gonadotrophin chain beta, Chorionic gonadotropin beta 3 subunit, chorionic gonadotropin beta chain, chorionic gonadotropin beta subunit 3, chorionic gonadotropin chain beta, hCGB, LHB, luteinizing hormone beta subunit
Host Species Rabbit
Immunogen Human embryonic kidney cell HEK293-derived transfected with human Chorionic Gonadotropin β, Accession # P0DN86 nad P0DN87
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1082
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.