Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ hCG beta-7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597102
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597102 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597102 Supplier Invitrogen™ Supplier No. PA597102
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SREMLRPRCR PINATLAVEK EGCPVCITVN TTICAGYCPT MTRVLQGVLP ALPQVVCNYR DVRFESIRLP GCPRGVNPVV SYAVALSCQC ALCRRSTTDC GGPKDHPLTC DDPRFQASSS SKAPPPSLPS PSRLPGPSDT PILPQ; Positive Samples: 293T, MCF-7, Mouse ovary, Mouse testis.

This gene is a member of the glycoprotein hormone beta chain family and encodes the beta 7 subunit of chorionic gonadotropin (CG). Glycoprotein hormones are heterodimers consisting of a common alpha subunit and an unique beta subunit which confers biological specificity. CG is produced by the trophoblastic cells of the placenta and stimulates the ovaries to synthesize the steroids that are essential for the maintenance of pregnancy. The beta subunit of CG is encoded by 6 genes which are arranged in tandem and inverted pairs on chromosome 19q13.3 and contiguous with the luteinizing hormone beta subunit gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen hCG beta-7
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.46 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CGB7
Gene Accession No. P0DN87
Gene Alias CGB6; CGB7; CG-beta-a; Choriogonadotropin subunit beta 7; chorionic gonadotropin beta 7 subunit; chorionic gonadotropin beta subunit 7; chorionic gonadotropin human; chorionic gonadotropin, beta polypeptide 7; human chorionic gonadotropin
Gene Symbols CGB7
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 21-165 of human CGB7 (NP_1491331).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 94027
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.