Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HCK Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144762
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144762 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144762 Supplier Invitrogen™ Supplier No. PA5144762
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CMKSKFLQVG GNTFSKTETS ASPHCPVYVP DPTSTIKPGP NSHNSNTPGI REAGSEDIIV VA Antigen sequence identity: Mouse 61%; Rat 60%.

Hematopoietic cell kinase (HCK) is a SRC-related kinase expressed in lymphoid and hematopoietic cells and may play a role in some forms of leukemia. (HCK Alias: DKFZp686D0638). HCK is a protein-tyrosine kinase that is predominantly expressed in hemopoietic cell types. The encoded protein may help couple the Fc receptor to the activation of the respiratory burst. In addition, it may play a role in neutrophil migration and in the degranulation of neutrophils. Alternate translation initiation site usage, including a non-AUG (CUG) codon, results in the production of two different isoforms, that have different subcellular localization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HCK
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HCK
Gene Accession No. P08631
Gene Alias AI849071; B cell/myeloid kinase; B-cell/myeloid kinase; BMK; HCK; HCK proto-oncogene, Src family tyrosine kinase; Hctk; Hematopoietic cell kinase; hemopoietic cell kinase; hemopoietic cell tyrosine kinase; JTK9; p56Hck; p56-HCK; p56-HCK/p59-HCK; p59Hck; p59-HCK/p60-HCK; p61Hck; tyrosine-protein kinase HCK
Gene Symbols HCK
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 24-85. Recombinant protein control fragment (Product #RP-109909).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3055
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.