Learn More
Invitrogen™ HCN2 Monoclonal Antibody (QQA 4A6), eBioscience™

Description
Description: This QQA 4A6 monoclonal antibody recognizes mouse and rat HCN2. QQA 4A6 detects a ~95 kDa band corresponding to HCN2 by Western Blot. An off target band of ~80 kDa is detected in cell line models. Applications Reported: This QQA 4A6 antibody has been reported for use in western blot and immunohistochemical staining. Applications Tested: This QQA 4A6 antibody has been tested by western blot and may be used at less than or equal to 1.0 μg/mL The QQA 4A6 antibody has been tested by immunohistochemistry of frozen mouse tissue sections and can be used at less than or equal to 40 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
Specifications
Specifications
| Antigen | HCN2 |
| Applications | Immunohistochemistry (Frozen), Western Blot |
| Classification | Monoclonal |
| Clone | QQA 4A6 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.2 |
| Gene | HCN2 |
| Gene Accession No. | O88703, Q9JKA9 |
| Gene Alias | BCNG2; BCNG-2; brain cyclic nucleotide gated channel 2; brain cyclic nucleotide-gated channel 2; Hac1; HAC-1; HCN2; hyperpolarization activated cyclic nucleotide gated potassium channel 2; hyperpolarization activated cyclic nucleotide-gated potassium channel 2; hyperpolarization-activated cation channel 1; hyperpolarization-activated, cyclic nucleotide-gated K+ 2; potassium/sodium hyperpolarization-activated cyclic nucleotide-gated channel 2; trls |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.