Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HCN3 Monoclonal Antibody (TLL6C5)
GREENER_CHOICE

Catalog No. MA3902 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA3902 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA3902 Supplier Invitrogen™ Supplier No. MA3902
  • $578.35
    $461.65 / Each of 1

    Save $116.70

    20% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rat Monoclonal Antibody

MA3-902 detects HCN3 from human, mouse and rat samples. MA3-902 has been successfully used in Western blotting and immunohistochemistry procedures. By Western blot MA3-902 detects a ~90 KDa band representing HCN3. The MA3-902 immunogen is a synthetic peptide corresponding to residues T(640) L L A R S A R R S A G S P A S P L V P V R A G P L L A R G P W A S T S(675) of rat HCN3. MA3-902 (clone TLL6C5) is a rat monoclonal hybridoma, produced by immunizing a rat, isolating the spleen cells, creating the hybridoma and successively injecting these cells into a mouse to produce ascites.

HCN3 is a member of the family of hyperpolarization-activated and cyclic-nucleotide-gated (HCN) channels. HCN currents have been linked to pacemaker activity in the heart and brain, resting potential control, as well as neuronal plasticity. HCN3 has been detected in the brain, but with less expression than other HCN channels.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HCN3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone TLL6C5
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Ascites with 0.05% sodium azide
Gene Hcn3
Gene Accession No. O88705, Q9JKA8, Q9P1Z3
Gene Alias Bcng4; BCNG-4; Hac3; HAC-3; HCN3; Hcn4; hyperpolarization activated cyclic nucleotide gated potassium channel 3; hyperpolarization activated cyclic nucleotide-gated potassium channel 3; hyperpolarization-activated cation channel 3; hyperpolarization-activated cyclic nucleotide-gated potassium channel 3; hyperpolarization-activated, cyclic nucleotide-gated K+ 3; hyperpolarization-activated, cyclic nucleotide-gated K+ 4; KIAA1535; MGC131493; Potassium/sodium hyperpolarization-activated cyclic nucleotide-gated channel 3
Gene Symbols Hcn3
Host Species Rat
Immunogen Synthetic peptide corresponding to residues T(640) L L A R S A R R S A G S P A S P L V P V R A G P L L A R G P W A S T S(675) of rat HCN3.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114245, 15168, 57657
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.