Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HCN4 Monoclonal Antibody (N114/10), PerCP
GREENER_CHOICE

Catalog No. PIMA545433
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA545433 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA545433 Supplier Invitrogen™ Supplier No. MA545433
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

1 μg/mL of MA5-45433 was sufficient for detection of HCN4 in 10 μg of COS cell lysate transiently transfected with HCN4 by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 130kDa. No cross-reactivity against other HCNs.

HCN4 is a member of the family of hyperpolarization-activated and cyclic-nucleotide-gated (HCN) channels. HCN currents have been linked to pacemaker activity in the heart and brain, resting potential control, as well as neuronal plasticity. It has been shown that HCN4 channels function as receptors for sour taste, and are associated with pacemaker potential generation in the sinoatrial node.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HCN4
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Immunohistochemistry (PFA fixed), Western Blot, Immunocytochemistry, Microarray
Classification Monoclonal
Clone N114/10
Concentration 1 mg/mL
Conjugate PerCP
Formulation 2.48mM MES, 95.64mM phosphate with 0.5M EDTA and no preservative; pH 7.4
Gene Hcn4
Gene Accession No. O70507, Q9JKA7, Q9TV66, Q9Y3Q4
Gene Alias Bcng3; BCNG-3; brain cyclic nucleotide-gated channel 3; Hcn3; Hcn4; hyperpolarization activated cyclic nucleotide gated potassium channel 4; hyperpolarization activated cyclic nucleotide-gated cation channel 4; hyperpolarization activated cyclic nucleotide-gated potassium channel 4; hyperpolarization-activated, cyclic nucleotide-gated K+ 3; hyperpolarization-activated, cyclic nucleotide-gated K+ 4; potassium/sodium hyperpolarization-activated cyclic nucleotide-gated channel 4; SSS2
Gene Symbols Hcn4
Host Species Mouse
Immunogen Fusion protein amino acids 1019-1198 (c-terminus) of rat HCN4.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100009452, 10021, 330953, 59266
Target Species Human, Mouse, Rat, Rabbit
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.