Learn More
Description
Peptide Sequence: LRMSEHRNEA LGNYLEMRLK SSFLRGLGSW KSNPLRLGGW TILLTLTMGQ GEPGGPQGDP WVPHELLLPS LCDSSHASSW GSGSITCAWR GGDSSSHPLV S.
Specifications
Specifications
| Antigen | HCP5 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 3G7 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | HCP5 |
| Gene Accession No. | Q6MZN7 |
| Gene Alias | 6S2650E; D6S2650E; GPHRB; HCP5; HLA complex P5 (non-protein coding); P5-1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
