Learn More
StatLab HCS Control Slides AFB Adhesion Slides
SDP
Catalog No. 23640162
Change view
Click to view available options
Quantity:
100/Pk.
25/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-640-162 100/Pk.
23-640-163 25/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-640-162 Supplier StatLab Supplier No. CS003PS100
Add to Cart
Add to Cart
HCS Control Slides AFB Adhesion Slides
  • HCS Control Slides consist of formalin fixed, paraffin embedded tissue sections which contain known positive tissue elements
  • Use these slides with confidence to evaluate stain quality and performance in histology laboratories
  • StatLab HCS control slides are created using adhesion white glass slides with 90° corners
  • Each package includes a specific staining protocol
Specifications
Type Control Slide
Quantity 100/Pk.