Learn More
ATCC HCT116-SLC15A4-KO-c12; Colorectal carcinoma; Human (Homo sapiens)
Supplier:  ATCC CRL3511
Encompass
The HCT116-SLC15A4-KO-c12 cell line contains a -19 deletion/-7 deletion in SLC15A4 gene and combined with the parental cell line and other HCT116 derivatives can be used in cancer drug screening.
Catalog No. 50-238-3120
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.