Learn More
Creative Biomart Recombinant HCV Core nucleocapsid Antigen, Biotin-Labled
Supplier:  Creative Biomart HCV155
Encompass_Preferred
The E. coli derived recombinant protein contains the HCV core nucleocapsid immunodominant regions, amino acids 2-192, 22 kDa.The Biotin labeled protein is fused with b-galactosidase (114 kDa) at N-ter
Catalog No. 50-208-4533
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.