Learn More
Creative Biomart Recombinant Chlamydia PGP-3D Antigen, His-tagged
Supplier:  Creative Biomart HCV178
Encompass_Preferred
The E. coli derived recombinant protein contains Chlamydia Trachomatis PGP-3D full length protein epitope. The protein is fused to a 6 His-Tag at C-terminus.
Catalog No. 50-208-4556
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.