Learn More
Creative Biomart Recombinant HCV Genotype 1b NS3 Antigen, GST-tagged
Supplier:  Creative Biomart HCV182
Encompass_Preferred
The E. coli derived recombinant protein contains the HCV NS3 immunodominant regions, amino acids 1356-1459. The protein is fused to a 22 kDa proprietary GST tag at N-Terminus.
Catalog No. 50-208-4560
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.