Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
- Home
- Products
- Research and Clinical Diagnostic Tests and Controls
- Chemistry Controls, Calibrators and Reagents
- General Microbiology Quality Control Products
- ZeptoMetrix HCV Seroconversion Panels - 3 to 10 Member Panel
ZeptoMetrix HCV Seroconversion Panels - 3 to 10 Member Panel
Catalog No. 22157215
Shop All ZeptoMetrix Corporation Products

Change view
Click to view available options
Quantity:
4 x 1 mL
5 x 1 mL
6 x 1 mL
7 x 1 mL
8 x 1 mL
9 x 1 mL
10 x 1 mL
Specimen Type:
Donor No.: 1038851
Donor No.: 22708D
Donor No.: 420245049
Donor No.: 490104584
Donor No.: 61025
Donor No.: 61251
Donor No.: 61404
Donor No.: 62258
Donor No.: 62286
Donor No.: 62680
Donor No.: 62804
Donor No.: 63318
Donor No.: 63625
Donor No.: 63792
Donor No.: 63869
Donor No.: 63911
Donor No.: 64150
Donor No.: 64254
Donor No.: 64273
Donor No.: 64437
Donor No.: 64766
Donor No.: 65470
Donor No.: 65643
Donor No.: 65977
Donor No.: 65982
Donor No.: 66074
Donor No.: 66626
Donor No.: 66647
Donor No.: 66719
Donor No.: 66751
Donor No.: 66822
Donor No.: 67116
Donor No.: 67356
Donor No.: 68157
Donor No.: 68629
Donor No.: 68896
Donor No.: 69028
Donor No.: 69089
Donor No.: 69142
Donor No.: 69263
Donor No.: 69654
Donor No.: 70225
Donor No.: 72237
Donor No.: 90163052
44 product options available for selection
| Catalog No. | Quantity | Specimen Type |
|---|---|---|
| 22-157-215 | 5 x 1 mL | Donor No.: 70225 |
| 22-157-176 | 6 x 1 mL | Donor No.: 69028 |
| 22-157-247 | 5 x 1 mL | Donor No.: 66647 |
| 22-157-233 | 8 x 1 mL | Donor No.: 62804 |
| 22-157-241 | 8 x 1 mL | Donor No.: 64437 |
| 22-157-226 | 8 x 1 mL | Donor No.: 62286 |
| 22-157-214 | 9 x 1 mL | Donor No.: 72237 |
| 22-157-204 | 10 x 1 mL | Donor No.: 65643 |
| 22-157-239 | 5 x 1 mL | Donor No.: 64254 |
| 22-157-183 | 10 x 1 mL | Donor No.: 67356 |
| 22-157-179 | 6 x 1 mL | Donor No.: 68896 |
| 22-157-210 | 7 x 1 mL | Donor No.: 420245049 |
| 22-157-244 | 10 x 1 mL | Donor No.: 66626 |
| 22-157-174 | 7 x 1 mL | Donor No.: 69654 |
| 22-157-205 | 4 x 1 mL | Donor No.: 65982 |
| 22-157-173 | 10 x 1 mL | Donor No.: 69089 |
| 22-157-192 | 8 x 1 mL | Donor No.: 66719 |
| 22-157-235 | 6 x 1 mL | Donor No.: 63792 |
| 22-157-228 | 6 x 1 mL | Donor No.: 62680 |
| 22-157-216 | 5 x 1 mL | Donor No.: 66822 |
| 22-157-212 | 8 x 1 mL | Donor No.: 490104584 |
| 22-157-187 | 8 x 1 mL | Donor No.: 69142 |
| 22-157-238 | 8 x 1 mL | Donor No.: 64150 |
| 22-157-225 | 8 x 1 mL | Donor No.: 62258 |
| 22-157-213 | 7 x 1 mL | Donor No.: 90163052 |
| 22-157-249 | 5 x 1 mL | Donor No.: 1038851 |
| 22-157-196 | 7 x 1 mL | Donor No.: 67116 |
| 22-157-248 | 5 x 1 mL | Donor No.: 22708D |
| 22-157-224 | 6 x 1 mL | Donor No.: 61404 |
| 22-157-221 | 5 x 1 mL | Donor No.: 61025 |
| 22-157-184 | 4 x 1 mL | Donor No.: 68157 |
| 22-157-231 | 7 x 1 mL | Donor No.: 63318 |
| 22-157-222 | 8 x 1 mL | Donor No.: 61251 |
| 22-157-180 | 7 x 1 mL | Donor No.: 65470 |
| 22-157-206 | 5 x 1 mL | Donor No.: 66074 |
| 22-157-178 | 10 x 1 mL | Donor No.: 69263 |
| 22-157-200 | 10 x 1 mL | Donor No.: 68629 |
| 22-157-240 | 10 x 1 mL | Donor No.: 64273 |
| 22-157-236 | 6 x 1 mL | Donor No.: 63869 |
| 22-157-195 | 8 x 1 mL | Donor No.: 66751 |
| 22-157-237 | 6 x 1 mL | Donor No.: 63911 |
| 22-157-234 | 8 x 1 mL | Donor No.: 63625 |
| 22-157-198 | 8 x 1 mL | Donor No.: 65977 |
| 22-157-242 | 5 x 1 mL | Donor No.: 64766 |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 44 options available.
Catalog No.
22-157-215
missing translation for 'mfr'
ZeptoMetrix
missing translation for 'supplierNo'
HCV10185
Add to Cart
Add to Cart
Specifications
Specifications
| Certifications/Compliance | FDA |
| Content And Storage | -20°C |
| Product Type | Member Panel |
| Quantity | 5 x 1 mL |
| Specimen Type | Donor No.: 70225 |
Product Title
Select an issue
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?