Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HDAC1 Mouse anti-Human, Mouse, Rat, Clone: 4C4G7, Proteintech
SDP

Catalog No. 501736522 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6522 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6522 Supplier Proteintech Group Inc Supplier No. 660851IG150UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

This antibody is raised against residues near the C terminus of human HDAC1. The calculated molecular weight of HDAC1 is 55 kDa, but modified HDAC1 is about 60-65 kDa.

Transcriptional coactivator for CREB1 which activates transcription through both consensus and variant cAMP response element (CRE) sites.Acts as a coactivator, in the SIK/TORC signaling pathway,being active when dephosphorylated and acts independently of CREB1 ′Ser-133′ phosphorylation.Enhances the interaction of CREB1 with TAF4.Regulates the expression of specific CREB-activated genes such as the steroidogenic gene,StAR.Potent coactivator of PGC1alpha and inducer of mitochondrial biogenesis in muscle cells.Also coactivator for TAX activation of the human T-cell leukemia virus type 1 (HTLV-1) long terminal repeats (LTR).In the hippocampus, involved in late-phase long-term potentiation (L-LTP) maintece at the Schaffer collateral-CA1 synapses.May be required for dendritic growth of developing cortical neurons.

Specifications

Antigen HDAC1
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 4C4G7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene HDAC1
Gene Accession No. O09106, Q13547, Q4QQW4
Gene Alias GON 10, HD1, HDAC1, histone deacetylase 1, RPD3, RPD3L1
Gene Symbols Hdac1
Host Species Mouse
Immunogen HDAC1 Fusion Protein Ag19128
Purification Method Protein G
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 297893, 3065, 433759
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.