Learn More
Invitrogen™ HDAC1 Polyclonal Antibody

Description
Suggested positive control: 293 whole cell lysate, antigen standard for HDAC1 (transient overexpression lysate).
Specifications
Specifications
| Antigen | HDAC1 |
| Applications | ChIP Assay, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | HDAC1 |
| Gene Accession No. | O09106, Q13547, Q4QQW4 |
| Gene Alias | DKFZp686H12203; GON-10; HD1; Hdac1; Hdac1-ps; Histone deacetylase 1; histone deacetylase 1-like protein; MommeD5; reduced potassium dependency, yeast homolog-like 1; RP4-811H24.2; RPD3; RPD3L1 |
| Gene Symbols | HDAC1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.