Learn More
Description
PA1-861 detects histone deacetylase 2 (HDAC2) from canine, hamster, human, mouse, non-human primate, rat tissues and cells. PA1-861 has been successfully used in Western blot, ChIP, immunoprecipitation, immunofluorescence, flow cytometry, immunocytochemistry and immunohistochemistry procedures. By Western blot, this antibody detects an ~55 kDa protein representing HDAC2 and a ~40 kDa protein of unknown identity from HeLa cell lysate. Immunocytochemical staining of HDAC2 in HeLa cells with PA1-861 results in nuclear staining. The PA1-861 immunogen is a synthetic peptide corresponding to residues C T(475) D T K G T K S E Q L S N P(488) of human HDAC2. This immunizing peptide (Cat. # PEP-070) is available for use in neutralization and control experiments.
Specifications
Specifications
| Antigen | HDAC2 |
| Applications | ChIP Assay, Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1mg/mL BSA and 0.05% sodium azide |
| Gene | HDAC2 |
| Gene Accession No. | P70288, Q92769 |
| Gene Alias | D10Wsu179e; HD2; hdac 2; Hdac2; hdac-2; Histone deacetylase 2; histone deacetylase 2-like protein; histone deacetylase-2; I79_010219; mRPD3; RPD3; transcriptional regulator homolog RPD3; YAF1; YY1 transcription factor-binding protein; YY1-associated factor 1; Yy1bp |
| Gene Symbols | HDAC2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
