Learn More
Invitrogen™ HDAC6 Polyclonal Antibody

Description
Suggested positive control: antigen standard for HDAC6 (transient overexpression lysate), NIH-3T3 whole cell lysate.
Specifications
Specifications
| Antigen | HDAC6 |
| Applications | ChIP Assay, Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | HDAC6 |
| Gene Accession No. | Q9UBN7, Q9Z2V5 |
| Gene Alias | CPBHM; FLJ16239; Hd6; HDA6; Hdac5; Hdac6; histone deacetylase 5; histone deacetylase 6; Histone deacetylase mHDA2; JM21; KIAA0901; mHDA2; PPP1R90; protein phosphatase 1, regulatory subunit 90; scurfy candidate 6; Sfc6 |
| Gene Symbols | HDAC6 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.