Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HDC Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558580
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558580 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558580 Supplier Invitrogen™ Supplier No. PA558580
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PALTSWPSLL GDMLADAINC LGFTWASSPA CTELEMNVMD WLAKMLGLPE HFLHHHPSSQ GGGVLQSTVS ESTLIALLAA RKNKILEMKT SEPDADESCL NARLVAYASD QAHSSVEK Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 94%.

The biogenic amine histamine is an important modulator of numerous physiologic processes, including neurotransmission, gastric acid secretion, and smooth muscle tone. The biosynthesis of histamine from histidine is catalyzed by the enzyme L-histidine decarboxylase (HDC; EC 4.1.1.22). This homodimeric enzyme is a pyridoxal phosphate (PLP)-dependent decarboxylase and is highly specific for its histidine substrate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HDC
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Hdc
Gene Accession No. P19113
Gene Alias AW108189; HDC; Hdc-a; Hdc-c; Hdc-e; Hdc-s; histidine decarboxylase; L-histidine decarboxylase; MGC163399
Gene Symbols Hdc
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HDC. Recombinant protein control fragment (Product #RP-88649).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3067
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.