Learn More
Medchemexpress LLC Hdl-16 | 2373280-36-3 | 99.2% | 303.15 g·mol^-1 | C14H11BrN2O | 10 MG

Supplier: Medchemexpress LLC HY16134310MG
HDL-16 is a small-molecule P2Y14 receptor antagonist used in research to study inflammation and necroptosis pathways. It is a brominated heterocyclic compound characterized by high potency in biochemical assays and supplied as a solid for laboratory applications.
- Potent P2Y14R antagonist with reported IC50 ≈ 0.3095 nM.
- Chemical formula C14H11BrN2O.
- Molecular weight 303.15 g·mol^-1.
- High purity (≈99% by LC/COA).
- Suitable for in vitro and preclinical research use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.