Learn More
Athens Research And Technology Inc HDL LIPOPROTEINS HU 1MG
Supplier:  Athens Research And Technology Inc 12160804121MG
Encompass_Preferred
Lipoproteins, High Density, Human Plasma: 1 MG; Liquid in 150 mM NaCl, pH 7.4, and 0.1% EDTA; MW 175,000-360,000. Native; Purified from fresh, non-frozen human plasma in the Athens Research & Tech lab. Molecular Biology Reagent, Lipoproteins
Catalog No. 50-489-856
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.