Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Justrite™ HDPE, Amber Carboy
GREENER_CHOICE

Catalog No. 19830421 Shop All Justrite Safety Group Products
Change view
Click to view available options
Capacity (Metric):
2.5 L
5 L
10 L
20 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
19-830-421 5 L
19-830-420 2.5 L
19-830-423 20 L
19-830-422 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-830-421 Supplier Justrite™ Supplier No. 12920
Add to Cart
Add to Cart

For Use With: Storing and dispensing solvents, buffers, reagents and powders in light sensitive applications; Closure Material: Polypropylene; Color: Amber

  • Interchangeable caps and adapters provide top connection flexibility
  • Rectangular ergonomic design
  • Material, volume, and metric graduation marks imprinted on carboys

Specifications

Capacity (English) 1.3 gal.
Capacity (Metric) 5 L
Closure Material Polypropylene
Closure Size 83 mm
Color Amber
Dimensions (L x W x H) 221 x 132 x 414 mm
For Use With (Application) Storing and dispensing solvents, buffers, reagents and powders in light sensitive applications
Height (English) 16.3 in.
Length (English) 8.7 in.
Material HDPE
Product Type Rectangular Carboy
Shape Rectangular
Width (English) 5.2 in.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.