Learn More
Description
- Durable HDPE design: Made from virgin-grade high-density polyethylene with a silicone-lined polypropylene cap, ideal for secure liquid storage in laboratories and industrial settings.
- Temperature range: Suitable for liquids between -100°C and +120°C, ensuring versatility in handling a variety of chemical and laboratory applications.
- FDA and USP 88 certified: Complies with FDA 21 CFR 177.1520 and USP 88 Class VI standards, making it safe for food, drug, and medical contact applications.
- Leakproof tested: Rigorously tested for shatter and crack resistance, handle strength, and leak prevention to ensure safe storage and transport of liquids.
- Multipurpose: Perfect for reagent storage, liquid mixing, and transportation in labs, research facilities, and educational settings. Features a stable base and ergonomic handles for ease of use.
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Capacity (Metric) | 20 L |
| Color | Translucent |
| Material | HDPE |
| Mouth Style | Narrow |
| Product Type | Carboy |
| Sterility | Non-sterile |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
