Learn More
Eisco Labs HDPE Carboy with Silicone-Lined Polypropylene Cap
SCI_ED
Catalog No. 1675234 Shop All Eisco Products
Change view
Click to view available options
Capacity (Metric):
20 L
50 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
16-752-34 50 L
16-752-33 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 16-752-34 Supplier Eisco Labs Supplier No. CH5565
Add to Cart
Add to Cart
HDPE carboy with a silicone-lined polypropylene cap are FDA and USP certified, leakproof, and chemically resistant, ideal for secure liquid storage and transport in lab, industrial, or educational environments.
  • Durable HDPE design: Made from virgin-grade high-density polyethylene with a silicone-lined polypropylene cap, ideal for secure liquid storage in laboratories and industrial settings.
  • Temperature range: Suitable for liquids between -100°C and +120°C, ensuring versatility in handling a variety of chemical and laboratory applications.
  • FDA and USP 88 certified: Complies with FDA 21 CFR 177.1520 and USP 88 Class VI standards, making it safe for food, drug, and medical contact applications.
  • Leakproof tested: Rigorously tested for shatter and crack resistance, handle strength, and leak prevention to ensure safe storage and transport of liquids.
  • Multipurpose: Perfect for reagent storage, liquid mixing, and transportation in labs, research facilities, and educational settings. Features a stable base and ergonomic handles for ease of use.
Specifications
Autoclavable Not Autoclavable
Capacity (Metric) 50 L
Color Translucent
Material HDPE
Mouth Style Narrow
Product Type Carboy
Sterility Non-sterile