Learn More
Description
- Interchangeable caps and adapters provide top connection flexibility
- Rectangular ergonomic design
- Material, volume, and metric graduation marks imprinted on carboys
Specifications
Specifications
| Capacity (English) | 10.6 gal. |
| Capacity (Metric) | 40 L |
| Closure Material | Polypropylene |
| Closure Size | 120 mm |
| Color | White |
| Dimensions (L x W x H) | 476 x 249 x 772 mm |
| For Use With (Application) | For storing and dispensing solvents, buffers, reagents and powders. |
| Height (English) | 30.4 in. |
| Length (English) | 18.75 in. |
| Material | HDPE |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.