Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Justrite™ HDPE, Carboy UN/DOT Approved
GREENER_CHOICE

Catalog No. 19830450 Shop All Justrite Safety Group Products
Change view
Click to view available options
Capacity (Metric):
13.5 L
20 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
19-830-450 13.5 L
19-830-451 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-830-450 Supplier Justrite™ Supplier No. 12950
Add to Cart
Add to Cart
This item is not returnable. View return policy

For Use With: Storage and secure shipments of liquid waste; Closure Material: HDPE; Color: White

  • For packaging group II and hazardous chemicals
  • HDPE construction for excellent chemical compatibility

Compliance

Meets UN 3H1 standards

Specifications

Capacity (English) 3.6 gal.
Capacity (Metric) 13.5 L
Certifications/Compliance UN/DOT
Closure Material HDPE
Closure Size 70 mm
Color White
Dimensions (L x W x H) 279 x 235 x 279 mm
For Use With (Application) Storage and secure shipments of liquid waste
Height (English) 11 in.
Length (English) 11 in.
Material HDPE
Product Type Carboy
Shape Rectangular
Width (English) 9.25 in.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.