Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ HE Filter TR-3712N

Catalog No. 19039579 Shop All 3M Products
Change view
Click to view available options
Quantity:
40/Cs.
5/Cs.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-039-579 40/Cs.
19-039-580 5/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-039-579 Supplier 3M™ Supplier No. 7100019023
Add to Cart
Add to Cart

HE Filter TR-3712N is a High Efficiency (HE) particulate filter for use with TR-300 Series PAPRs.

  • High Efficiency Particulate Filter
  • Convenient package sizes
  • 3M Catalog# TR-3712N-40 , 3M Catalog# TR-3712N-5

TRUSTED_SUSTAINABILITY

Specifications

Color Magenta
Product Type HE Particulate Filter
For Use With (Equipment) TR-300 Series PAPRs
Model TR-3712N-40
Quantity 40/Cs.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.