Learn More
Description
- High Efficiency Particulate Filter
- Convenient package sizes
- 3M Catalog# TR-3712N-40 , 3M Catalog# TR-3712N-5
Specifications
Specifications
| Color | Magenta |
| Product Type | HE Particulate Filter |
| For Use With (Equipment) | TR-300 Series PAPRs |
| Model | TR-3712N-5 |
| Quantity | 5/Cs. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.