Learn More
Apexbio Technology LLC Hematoxylin and Eosin Staining Kit 500mL
SDP
Supplier:  Apexbio Technology LLC K1142500
Encompass_Preferred
Hematoxylin and Eosin (H E) staining is commonly employed to visualize tissue morphology in histopathological analysis Hematoxylin functions by oxidation and subsequent complex formation with metal mordants (e g aluminum or iron salts) yielding positively charged dye complexes that selectively bind to negatively charged phosphates within cell nuclei producing a blue or bluish-purple coloration Eosin an acidic dye with negatively charged ions stains cytoplasmic components and extracellular matrix proteins pink or reddish through electrostatic interactions with positively charged amino groups This method is extensively used for assessing cellular structures and tissue pathology in paraffin or frozen tissue sections as well as cytological preparations Provided solutions are ready-to-use working concentrations compatible for direct staining protocols without further dilution
Catalog No. 50-000-63339
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.