Learn More
CPC Scientific Glp-Pro-Pro-Gly-Gly-Ser-Lys-Val-Ile-Leu-Phe-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific NEUP112A
Encompass
Morphogenic peptide from hydra.ONE-LETTER SEQUENCE: Glp-PPGGSKVILFMOLECULAR FORMULA: C54H84N12O14MOLECULAR WEIGHT:1125.33STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [79943-68-3], SYNONYMS: Head activatorRESEARCH AREA: NeuropeptidesREFERENCES: H. Bodenmuller et al., Nature, 293, 579 (1981)
Catalog No. 50-210-9943
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.