Learn More
ATCC Head and Neck Cancer Cell Line Panel
Supplier:  ATCC TCP1012
Encompass
A panel of 6 cell lines from salivary gland, tongue, hypopharynx and pharynx. Each culture contains genomic mutations in one or more of the following genes according to the Sanger COSMIC database: CDKN2A, TP53, SMAD4, PIK3CA and KDM5C.
Catalog No. NC2684004