Learn More
PIP™ Headgear for Face Protection with Ratchet Suspension
Catalog No. 19046413 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Blue
Catalog No.
19-046-413 Blue
1 options
This item is not returnable. View return policy
Catalog No. 19-046-413 Supplier PIP™ Supplier No. 251015200
This item is not returnable. View return policy
PIP™ Headgear provides crown protection in grinding applications. Visors are available separately.
  • Ratchet suspension provides effortless size adjustment
  • Lightweight, durable blue polypropylene
  • Premium fine tooth ratchet suspension offers more control for an exact fit
  • Visor attaches easily with pin adjustment, and conveniently flips up out of the way when not in use
  • Includes brow sweatband
  • Meets ANSI Z87.1+
  • Meets CSA Z94.3
Specifications
For Use With (Application) Face protection
Frame Color Blue