Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Heartland Virus Glycoprotein 1 Synthetic Peptide

Catalog No. PEP1618
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP1618 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP1618 Supplier Invitrogen™ Supplier No. PEP1618
Add to Cart
Add to Cart

Synthetic Peptide

Heartland Virus Glycoprotein 1 Synthetic Peptide, PEP-1618, from Invitrogen.

The Heartland virus is a newly identified member of the phlebovirus genus of the Bunyaviridae family. Initial reports indicate that symptoms may include fever, fatigue, diarrhea, thrombocytopenia, and leukopenia. The Bunyaviridae family of viruses are spherical enveloped viruses with a tripartite RNA genome of negative or ambisense polarity. The glycoprotein RNA encodes a polyprotein that is cleaved into the G1 and G2 proteins.
TRUSTED_SUSTAINABILITY

Specifications

Species Virus
Purification Method purified
Common Name Heartland Virus Glycoprotein 1
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Lyophilized No
Preservative 0.02% sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.