Learn More
PHILIPS MEDICAL SYSTEMS HEARTSTART BATTERY PACK
Supplier:  PHILIPS MEDICAL SYSTEMS 989803121381
Encompass
HeartStart Onsite And FRX Defibrillator Battery for use with FRx/OnSite/AED Defibrillator. Long-life lithium battery comes with 4-year factory warranty. A spare battery should be stored with the defibrillator.
Catalog No. NC9767160
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.